Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzhydryl-3-azetidinyl methanesulfonate 97%

Supplier: AMBEED, INC
Warning

A288967-10G A288967-1G A288967-25G A288967-5G
77032-262EA 15 USD InStock
77032-262 77032-264 77032-266 77032-268
1-Benzhydryl-3-azetidinyl methanesulfonate 97%
1-Benzhydryl-3-azetidinyl methanesulfonate
Precaución: for Research Use Only


Formula: C₁₇H₁₉NO₃S
MDL Number: MFCD00159216
CAS Number: 33301-41-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty