Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxy-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: AMBEED, INC
Warning

A289298-1G A289298-250MG A289298-25G A289298-5G
77014-734EA 65 USD InStock
77014-734 77014-736 77014-738 77014-740
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline 97%
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₁₁NO
MDL Number: MFCD01717047
CAS Number: 14446-24-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty