Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-2-methoxypyridine 95+%

Supplier: AMBEED, INC
Warning

A387456-1G A387456-250MG A387456-5G
77016-138EA 105 USD InStock
77016-138 77016-140 77016-142
5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-2-methoxypyridine 95+%
5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-2-methoxypyridine
Precaución: for Research Use Only


Formula: C₁₁H₁₆BNO₃
MDL Number: MFCD06801680
CAS Number: 1022094-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty