Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-trans-cyclohexane-1,2-dicarboxylate 96%

Supplier: AMBEED, INC
Warning

A392902-100MG A392902-1G A392902-250MG A392902-5G
76990-156EA 5 USD InStock
76990-156 76990-158 76990-160 76990-162
Dimethyl-trans-cyclohexane-1,2-dicarboxylate 96%
Dimethyl trans-cyclohexane-1,2-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₆O₄
MDL Number: MFCD09998658
CAS Number: 3205-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty