Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole 96%

Supplier: AMBEED, INC
Warning

A463807-100MG A463807-1G A463807-250MG A463807-25G A463807-5G
77034-216EA 13 USD InStock
77034-216 77034-218 77034-220 77034-222 77034-224
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole 96%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole
Precaución: for Research Use Only


Formula: C₁₄H₁₈BNO₃
MW: 259.11 g/mol
MDL Number: MFCD08669935
CAS Number: 837392-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty