Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitropyridine-2-carbonitrile 98%

Supplier: AMBEED, INC
Danger

A473603-1G A473603-250MG A473603-25G A473603-5G
77016-946EA 55 USD InStock
77016-946 77016-948 77016-950 77016-952
4-Nitropyridine-2-carbonitrile 98%
4-Nitropyridine-2-carbonitrile
Precaución: for Research Use Only


Formula: C₆H₃N₃O₂
MDL Number: MFCD00191627
CAS Number: 19235-88-2
UN: 2923
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty