Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Bromoquinolin-2(1H)-one 98%

Supplier: AMBEED, INC
Warning

A496132-100MG A496132-10G A496132-250MG A496132-5G A496132-1G
77034-452EA 6 USD InStock
77034-452 77034-454 77034-456 77034-458 77669-224
8-Bromoquinolin-2(1H)-one 98%
8-Bromoquinolin-2(1H)-one
Precaución: for Research Use Only


Formula: C₉H₆BrNO
MDL Number: MFCD08741409
CAS Number: 67805-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty