Nickel(II) trifluoromethanesulfonate 98%

Supplier: AMBEED, INC
Danger

A527892-25G A527892-5G
77034-706EA 42 USD
77034-706 77034-708
Nickel(II) trifluoromethanesulfonate 98%
Nickel(II) trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂F₆NiO₆S₂
MW: 356.82 g/mol
MDL Number: MFCD00673740
CAS Number: 60871-84-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR