Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Macitentan 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A539182-10MG A539182-1MG A539182-50MG A539182-5MG A539182-100MG A539182-5G
77017-564EA 9 USD InStock
77017-564 77017-566 77017-568 77017-570 77613-690 77684-380
Macitentan 98%, Ambeed.Inc
Macitentan
Precaución: For research use only.


Formula: C₁₉H₂₀Br₂N₆O₄S
MDL Number: MFCD17167076
CAS Number: 441798-33-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty