Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-(S)-2-Cyclobutylglycine 98%

Supplier: AMBEED, INC
Warning

A552906-100MG A552906-1G A552906-250MG
77005-098EA 24 USD InStock
77005-098 77005-100 77005-102
Boc-(S)-2-Cyclobutylglycine 98%
Boc-(S)-2-Cyclobutylglycine
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
MDL Number: MFCD06659975
CAS Number: 155905-77-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty