Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Oxiran-2-ylmethyl 4-nitrobenzenesulfonate 98+%

Supplier: AMBEED, INC
Warning

A635502-100G A635502-1G A635502-25G A635502-5G
77005-628EA 700 USD InStock
77005-628 77005-630 77005-632 77005-634
(R)-Oxiran-2-ylmethyl 4-nitrobenzenesulfonate 98+%
(R)-Oxiran-2-ylmethyl 4-nitrobenzenesulfonate
Precaución: for Research Use Only


Formula: C₉H₉NO₆S
MDL Number: MFCD08460169
CAS Number: 123750-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty