Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylphenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A658676-100G A658676-10G A658676-1G A658676-25G A658676-5G
77018-356EA 864 USD InStock
77018-356 77018-358 77018-360 77018-362 77018-364
4-Ethylphenylboronic acid pinacol ester 98%
4-Ethylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₁BO₂
MDL Number: MFCD18643353
CAS Number: 1075719-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty