Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-(Ethane-1,2-diyldisulfonyl)diethanol 95%

Supplier: AMBEED, INC
Warning

A672347-1G A672347-250MG
77018-478EA 140 USD InStock
77018-478 77018-480
2,2'-(Ethane-1,2-diyldisulfonyl)diethanol 95%
2,2'-(Ethane-1,2-diyldisulfonyl)diethanol
Precaución: for Research Use Only


Formula: C₆H₁₄O₆S₂
MDL Number: MFCD27923650
CAS Number: 7484-34-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty