Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(2-aminothiazol-4-yl)benzoate 98%

Supplier: AMBEED, INC
Warning

A681895-100MG A681895-1G A681895-250MG A681895-5G
77005-886EA 10 USD InStock
77005-886 77005-888 77005-890 77005-892
Methyl-4-(2-aminothiazol-4-yl)benzoate 98%
Methyl-4-(2-aminothiazol-4-yl)benzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂O₂S
MDL Number: MFCD03030162
CAS Number: 206555-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty