Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-indoleboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A690622-1G A690622-250MG A690622-5G
77018-664EA 210 USD InStock
77018-664 77018-666 77018-668
6-Bromo-2-indoleboronic acid pinacol ester 98%
6-Bromo-2-indoleboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₇BBrNO₂
MDL Number: MFCD11858393
CAS Number: 1218791-00-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty