Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Fluoro-3'-hydroxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A722479-100MG A722479-1G A722479-250MG A722479-5G
76992-858EA 11 USD InStock
76992-858 76992-860 76992-862 76992-864
4'-Fluoro-3'-hydroxyacetophenone 98%
4'-Fluoro-3'-hydroxyacetophenone
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MDL Number: MFCD12545938
CAS Number: 949159-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty