4-Chloro-2-[(methylamino)methyl]phenol 95%
Supplier: AMBEED, INC
|
Warning
|
A741762-250MG
A741762-5G
77018-958EA
13.8
USD
77018-958
77018-960
77018-962
4-Chloro-2-[(methylamino)methyl]phenol 95%
4-Chloro-2-[(methylamino)methyl]phenol
Precaución: for Research Use Only
Formula:
C₈H₁₀ClNO MW: 171.624 g/mol |
MDL Number:
MFCD09044328 CAS Number: 38926-77-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...