Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-N-methylaniline hydrochloride 98%

Supplier: AMBEED, INC
Warning

A742522-10G A742522-1G A742522-25G A742522-5G
76992-970EA 34 USD InStock
76992-970 76992-972 76992-974 76992-976
4-Methoxy-N-methylaniline hydrochloride 98%
4-Methoxy-N-methylaniline hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₂ClNO
MW: 173.6400 g/mol
MDL Number: MFCD13196471
CAS Number: 10541-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty