Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl pyrrolo[1,2-b]pyridazine-6-carboxylate 95%

Supplier: AMBEED, INC
Warning

A783332-100MG A783332-250MG
76993-196EA 5 USD InStock
76993-196 76993-198
Methyl pyrrolo[1,2-b]pyridazine-6-carboxylate 95%
Methyl pyrrolo[1,2-b]pyridazine-6-carboxylate
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
MDL Number: MFCD12924819
CAS Number: 108128-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty