Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dibromo-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: AMBEED, INC
Warning

A868081-100MG A868081-1G A868081-250MG
77019-776EA 18 USD InStock
77019-776 77019-778 77019-780
3,6-Dibromo-1H-pyrrolo[3,2-c]pyridine 95%
3,6-Dibromo-1H-pyrrolo[3,2-c]pyridine
Precaución: for Research Use Only


Formula: C₇H₄Br₂N₂
MDL Number: MFCD09749985
CAS Number: 1000341-69-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty