3-Bromo-imidazo[1,2-a]pyridine-8-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A923482-100MG A923482-1G A923482-250MG
76993-896EA 31 USD
76993-896 76993-898 76993-900
3-Bromo-imidazo[1,2-a]pyridine-8-carboxylic acid 95%
3-Bromo-imidazo[1,2-a]pyridine-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.05 g/mol
MDL Number: MFCD17016090
CAS Number: 1116691-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR