Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Chlorophenyl)[1,3,4]thiadiazol-2-ylamine 95%

Supplier: AMBEED, INC
Warning

A996851-1G A996851-250MG A996851-5G
77007-294EA 41 USD InStock
77007-294 77007-296 77007-298
5-(4-Chlorophenyl)[1,3,4]thiadiazol-2-ylamine 95%
5-(4-Chlorophenyl)[1,3,4]thiadiazol-2-ylamine
Precaución: for Research Use Only


Formula: C₈H₆ClN₃S
MDL Number: MFCD00980966
CAS Number: 28004-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty