Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-methanesulfonylcinnamate 98%

Supplier: AMBEED, INC
Warning

A155447-100MG A155447-1G A155447-250MG
77095-944EA 28 USD InStock
77095-944 77095-946 77095-948
Ethyl 4-methanesulfonylcinnamate 98%
Ethyl 4-methanesulfonylcinnamate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₄S
MDL Number: MFCD22124828
CAS Number: 137473-27-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty