Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-4-(2-propoxybenzamido)-3-propyl-1H-pyrazole-5-carboxamide 97%

Supplier: AMBEED, INC
Warning

A197271-1G A197271-250MG A197271-25G A197271-5G
77097-286EA 7 USD InStock
77097-286 77097-288 77097-290 77097-292
1-Methyl-4-(2-propoxybenzamido)-3-propyl-1H-pyrazole-5-carboxamide 97%
1-Methyl-4-(2-propoxybenzamido)-3-propyl-1H-pyrazole-5-carboxamide
Precaución: for Research Use Only


Formula: C₁₈H₂₄N₄O₃
MDL Number: MFCD20270864
CAS Number: 139756-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty