Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

c-Met inhibitor 1 ≥98%

Supplier: AMBEED, INC
Warning

A248902-10MG A248902-1MG A248902-2MG A248902-50MG A248902-5MG
77098-632EA 52 USD InStock
77098-632 77098-634 77098-636 77098-638 77098-640
c-Met inhibitor 1 ≥98%
c-Met inhibitor 1
Precaución: for Research Use Only


Formula: C₁₇H₁₄N₈S
MDL Number: MFCD26960936
CAS Number: 1357072-61-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty