Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AT7519 hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A353288-10MG A353288-1MG A353288-50MG A353288-5MG
77106-806EA 74 USD InStock
77106-806 77106-808 77106-810 77106-812
AT7519 hydrochloride ≥98%
AT7519 hydrochloride
Precaución: for Research Use Only


Formula: C₁₆H₁₇Cl₂N₅O₂·HCl
MDL Number: MFCD14636428
CAS Number: 902135-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty