Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6,7-Trimethoxy-1H-benzo[d][1,3]oxazine-2,4-dione 95%

Supplier: AMBEED, INC
Warning

A413805-100MG A413805-1G A413805-250MG
77102-060EA 70 USD InStock
77102-060 77102-062 77102-064
5,6,7-Trimethoxy-1H-benzo[d][1,3]oxazine-2,4-dione 95%
5,6,7-Trimethoxy-1H-benzo[d][1,3]oxazine-2,4-dione
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₆
MDL Number: MFCD19109821
CAS Number: 124346-74-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty