Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylic acid 96%

Supplier: AMBEED, INC
Warning

A600869-1G A600869-250MG A600869-5G
77104-028EA 143 USD InStock
77104-028 77104-030 77104-032
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylic acid 96%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅BO₅
MDL Number: MFCD18383001
CAS Number: 1220188-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty