Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Dimethyl 2-((tert-butoxycarbonyl)amino)pentanedioate 97%

Supplier: AMBEED, INC
Warning

A939400-100G A939400-25G A939400-5G
77106-306EA 143 USD InStock
77106-306 77106-308 77106-310
(R)-Dimethyl 2-((tert-butoxycarbonyl)amino)pentanedioate 97%
(R)-Dimethyl 2-((tert-butoxycarbonyl)amino)pentanedioate
Precaución: for Research Use Only


Formula: C₁₂H₂₁NO₆
MDL Number: MFCD03094773
CAS Number: 130622-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty