(S)-(+)-3-Hydroxytetrahydrofuran 97%
Supplier: AMBEED, INC
|
Warning
|
A124923-100G
A124923-10G
A124923-1G
A124923-25G
A124923-500G
A124923-5G
77108-788EA
92.7
USD
77108-788
77108-790
77108-792
77108-794
77108-796
77108-798
(S)-(+)-3-Hydroxytetrahydrofuran 97%
(S)-(+)-3-Hydroxytetrahydrofuran
Precaución: for Research Use Only
Formula:
C₄H₈O₂ MW: 88.11 g/mol |
MDL Number:
MFCD00064327 CAS Number: 86087-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...