Iguratimod 98%

Supplier: AMBEED, INC
Warning

A178057-100MG A178057-10MG A178057-1G A178057-1MG A178057-250MG A178057-25G A178057-50MG A178057-5MG A178057-5G
77111-284EA 17.52 USD
77111-284 77111-286 77111-288 77111-290 77111-292 77111-294 77111-296 77111-298 77685-401
Iguratimod 98%
Iguratimod

Precaución: for Research Use Only

Formula: C₁₇H₁₄N₂O₆S
MW: 374.37 g/mol
MDL Number: MFCD00882374
CAS Number: 123663-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR