Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-2-fluorophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A464605-100MG A464605-1G A464605-250MG A464605-5G
77116-322EA 24.73 USD InStock
77116-322 77116-324 77116-326 77116-328
4-Cyano-2-fluorophenylboronic acid 98%
4-Cyano-2-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₅BFNO₂
MDL Number: MFCD07644472
CAS Number: 1150114-77-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty