Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(4-formylphenyl) ether 98%

Supplier: AMBEED, INC
Warning

A475772-1G A475772-250MG A475772-5G
77116-394EA 29.87 USD InStock
77116-394 77116-396 77116-398
Bis(4-formylphenyl) ether 98%
Bis(4-formylphenyl) ether
Precaución: for Research Use Only


Formula: C₁₄H₁₀O₃
MDL Number: MFCD08276812
CAS Number: 2215-76-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty