Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyindoline hydrochloride 95%

Supplier: AMBEED, INC
Warning

A572374-100MG A572374-1G A572374-250MG A572374-5G
77117-112EA 58.72 USD InStock
77117-112 77117-114 77117-116 77117-118
4-Methoxyindoline hydrochloride 95%
4-Methoxyindoline hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClNO
MDL Number: MFCD09026828
CAS Number: 90609-70-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty