Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Trifluoromethylphenyl)pyrrolidine 95%

Supplier: AMBEED, INC
Warning

A700501-1G A700501-250MG A700501-5G
77117-920EA 92.7 USD InStock
77117-920 77117-922 77117-924
1-(4-Trifluoromethylphenyl)pyrrolidine 95%
1-(4-Trifluoromethylphenyl)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₁H₁₂F₃N
MDL Number: MFCD16140176
CAS Number: 113845-68-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty