Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SAMe-1,4-Butanedisulfonate 95%

Supplier: AMBEED, INC
Warning

A110093-10G A110093-25G A110093-250MG
77135-956EA 139.05 USD InStock
77135-956 77135-958 77745-819
SAMe-1,4-Butanedisulfonate 95%
SAMe-1,4-Butanedisulfonate
Precaución: for Research Use Only


Formula: C₄₂H₇₄N₁₂O₂₈S₈
MDL Number: MFCD11111819
CAS Number: 101020-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty