Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-(3-Fluorophenyl)-5-(hydroxymethyl)oxazolidin-2-one 97%

Supplier: AMBEED, INC
Warning

A275598-1G A275598-250MG A275598-5G A275598-100MG
77141-040EA 63.86 USD InStock
77141-040 77141-042 77141-044 77746-491
(R)-3-(3-Fluorophenyl)-5-(hydroxymethyl)oxazolidin-2-one 97%
(R)-3-(3-Fluorophenyl)-5-(hydroxymethyl)oxazolidin-2-one
Precaución: for Research Use Only


Formula: C₁₀H₁₀FNO₃
MDL Number: MFCD08234640
CAS Number: 149524-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty