Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Difluoro-2-iodobenzene 98%

Supplier: AMBEED, INC
Warning

A281084-100G A281084-10G A281084-25G A281084-500G A281084-5G
77141-166EA 78.28 USD InStock
77141-166 77141-168 77141-170 77141-172 77141-174
1,3-Difluoro-2-iodobenzene 98%
1,3-Difluoro-2-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₃F₂I
MDL Number: MFCD04039247
CAS Number: 13697-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty