Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Dimethyl-3-(4-(trifluoromethyl)phenyl)pyrimido[5,4-e][1,2,4]triazine-5,7(1H,6H)-dione 98%

Supplier: AMBEED, INC
Warning

A299036-10MG A299036-1MG A299036-25MG A299036-50MG A299036-5MG
77141-504EA 125.66 USD InStock
77141-504 77141-506 77141-508 77141-510 77141-512
1,6-Dimethyl-3-(4-(trifluoromethyl)phenyl)pyrimido[5,4-e][1,2,4]triazine-5,7(1H,6H)-dione 98%
1,6-Dimethyl-3-(4-(trifluoromethyl)phenyl)pyrimido[5,4-e][1,2,4]triazine-5,7(1H,6H)-dione
Precaución: for Research Use Only


Formula: C₁₄H₁₀F₃N₅O₂
MDL Number: MFCD07188899
CAS Number: 878419-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty