Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chloroquinazolin-2-yl)-N,N-dimethylaniline 97%

Supplier: AMBEED, INC
Warning

A384179-100MG A384179-1G A384179-250MG
77142-624EA 21.63 USD InStock
77142-624 77142-626 77142-628
4-(4-Chloroquinazolin-2-yl)-N,N-dimethylaniline 97%
4-(4-Chloroquinazolin-2-yl)-N,N-dimethylaniline
Precaución: for Research Use Only


Formula: C₁₆H₁₄ClN₃
MDL Number: MFCD13464749
CAS Number: 79916-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty