Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-2-propyl Dodecyl trithiocarbonate 97%

Supplier: AMBEED, INC
Warning

A407164-250MG A407164-1G
77143-006EA 18.54 USD InStock
77143-006 77551-222
2-Cyano-2-propyl Dodecyl trithiocarbonate 97%
2-Cyano-2-propyl Dodecyl trithiocarbonate
Precaución: for Research Use Only


Formula: C₁₇H₃₁NS₃
MDL Number: MFCD11973800
CAS Number: 870196-83-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty