Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methylsulfonyl)pyridin-2(1H)-one 97%

Supplier: AMBEED, INC
Warning

A472583-100MG A472583-1G A472583-250MG
77143-694EA 122.57 USD InStock
77143-694 77143-696 77143-698
4-(Methylsulfonyl)pyridin-2(1H)-one 97%
4-(Methylsulfonyl)pyridin-2(1H)-one
Precaución: for Research Use Only


Formula: C₆H₇NO₃S
MDL Number: MFCD18632643
CAS Number: 1268519-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty