Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-3-(chloromethyl)-1H-indazole 97%

Supplier: AMBEED, INC
Danger

A550759-100MG A550759-250MG
77144-484EA 96.82 USD InStock
77144-484 77144-486
5-Chloro-3-(chloromethyl)-1H-indazole 97%
5-Chloro-3-(chloromethyl)-1H-indazole
Precaución: for Research Use Only


Formula: C₈H₆Cl₂N₂
MDL Number: MFCD24458996
CAS Number: 27328-69-4
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty