Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Nitropropiophenone 98%

Supplier: AMBEED, INC
Warning

A154713-100G A154713-10G A154713-1G A154713-250MG A154713-25G
77150-606EA 637 USD InStock
77150-606 77150-608 77150-610 77150-612 77150-614
3'-Nitropropiophenone 98%
3'-Nitropropiophenone
Precaución: for Research Use Only


Formula: C₉H₉NO₃
MDL Number: MFCD00051516
CAS Number: 17408-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty