Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide 95%

Supplier: AMBEED, INC
Warning

A183186-100G A183186-10G A183186-25G
77151-674EA 40 USD InStock
77151-674 77151-676 77151-678
N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide 95%
N-Ethyl-2-isopropyl-5-methylcyclohexanecarboxamide
Precaución: for Research Use Only


Formula: C₁₃H₂₅NO
MDL Number: MFCD00130071
CAS Number: 39711-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty