Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

WZ 811 98%

Supplier: AMBEED, INC
Warning

A220173-100MG A220173-10MG A220173-1MG A220173-50MG A220173-5MG
77152-670EA 51 USD InStock
77152-670 77152-672 77152-674 77152-676 77152-678
WZ 811 98%
WZ 811
Precaución: for Research Use Only


Formula: C₁₈H₁₈N₄
MDL Number: MFCD18086914
CAS Number: 55778-02-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty