Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,7-Trihydroxy-2,8-dimethoxy-6-methylanthracene-9,10-dione 98%

Supplier: AMBEED, INC
Warning

A221042-100MG A221042-250MG A221042-50MG
77152-684EA 170 USD InStock
77152-684 77152-686 77152-688
1,3,7-Trihydroxy-2,8-dimethoxy-6-methylanthracene-9,10-dione 98%
1,3,7-Trihydroxy-2,8-dimethoxy-6-methylanthracene-9,10-dione
Precaución: for Research Use Only


Formula: C₁₇H₁₄O₇
MDL Number: MFCD13194886
CAS Number: 67979-25-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty