Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Aminononanoic acid 98+%

Supplier: AMBEED, INC
Warning

A260748-1G A260748-250MG
77153-458EA 54 USD InStock
77153-458 77153-460
(S)-2-Aminononanoic acid 98+%
(S)-2-Aminononanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₉NO₂
MDL Number: MFCD08275755
CAS Number: 133444-84-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty