Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Cbz-4-piperidinecarboxylic acid methyl ester 97%

Supplier: AMBEED, INC
Warning

A320217-100G A320217-10G A320217-25G A320217-500G
77154-496EA 43 USD InStock
77154-496 77154-498 77154-500 77154-502
N-Cbz-4-piperidinecarboxylic acid methyl ester 97%
N-Cbz-4-piperidinecarboxylic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO₄
MW: 277.316 g/mol
MDL Number: MFCD06203777
CAS Number: 138163-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty