Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3,4,5-trifluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A342600-100G A342600-25G
77154-882EA 101 USD InStock
77154-882 77154-884
Ethyl 3,4,5-trifluorobenzoate 97%
Ethyl 3,4,5-trifluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₇F₃O₂
MDL Number: MFCD06204428
CAS Number: 495405-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty